siRNA Details
| Gene Symbol: PRODH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195226.1 | NP_001182155.1 | 2 | 127 | ||
| NM_016335.4 | NP_057419.4 | 3 | 542 | |||
| GenBank mRNA | AB209472.1 | - | 2 | 193 | ||
| BC036534.1 | - | 2 | 351 | |||
| BC068260.1 | - | 2 | 344 | |||
| BC094736.1 | - | 2 | 172 | |||
| BC118597.1 | - | 2 | 127 | 40108132 | ||
| BC121809.1 | - | 2 | 127 | 40108133 | ||
| CN362765.1 | - | 2 | 293 | |||
| KJ534920.1 | - | 2 | 338 | |||
| U79754.1 | - | 3 | 542 | |||
| Gene Symbol: LOC102724788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006724935.3 | XP_006724998.1 | 2 | 371 | ||
| XM_006724936.3 | XP_006724999.1 | 2 | 346 | |||
| XM_017030166.1 | XP_016885655.1 | 2 | 468 | |||
| XM_017030168.1 | XP_016885657.1 | 2 | 189 | |||

PRODH
Genomic Map