siRNA Details
Gene Symbol: GPR107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136557.1 | NP_001130029.1 | 1, 2 | 376 | ||
NM_001136558.1 | NP_001130030.1 | 1, 2 | 376 | |||
NM_001287346.1 | NP_001274275.1 | 1, 2 | 376 | |||
NM_020960.4 | NP_066011.2 | 1, 2 | 376 | |||
XM_017014953.1 | XP_016870442.1 | 1, 2 | 337 | |||
XM_017014954.1 | XP_016870443.1 | 1, 2 | 337 | |||
GenBank mRNA | AB046844.1 | - | 1, 2 | 147 | ||
AF376725.1 | - | 1, 2 | 157 | |||
AK026244.1 | - | 1, 2 | 161 | |||
AK225456.1 | - | 1, 2 | 161 | |||
AK293092.1 | - | 1, 2 | 157 | |||
AL834359.1 | - | 1 | 20 | |||
BC110518.2 | - | 1, 2 | 157 | 40033360 | ||
BC143655.1 | - | 1, 2 | 172 | |||
BM920552.1 | - | 375 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
