siRNA Details
| Gene Symbol: C1orf198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136494.1 | NP_001129966.1 | 3 | 232 | ||
| NM_032800.2 | NP_116189.1 | 1 | 174 | |||
| GenBank mRNA | AK027431.1 | - | 1 | 149 | ||
| AK096166.1 | - | 3 | 232 | |||
| BC066649.1 | - | 1 | 162 | 6011914 | ||

C1orf198
Genomic Map