siRNA Details
| Gene Symbol: CCDC122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144974.3 | NP_659411.2 | 4 | 405 | ||
| XM_017020397.1 | XP_016875886.1 | 4 | 523 | |||
| XM_017020398.1 | XP_016875887.1 | 4 | 523 | |||
| XM_017020399.1 | XP_016875888.1 | 4 | 523 | |||
| GenBank mRNA | AK056408.1 | - | 4 | 406 | ||
| BC137289.1 | - | 3 | 325 | 9021286 | ||
| BC137290.1 | - | 3 | 325 | 9021287 | ||
| BC144570.1 | - | 3 | 325 | 9053098 | ||
| BC144571.1 | - | 3 | 325 | 9053099 | ||
| DA739121.1 | - | 4 | 406 | |||

CCDC122
Genomic Map