siRNA Details
| Gene Symbol: HMGB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145205.5 | NP_660206.2 | 2 | 1869 | ||
| NR_033264.1 | 2 | 154 | ||||
| GenBank mRNA | AK097707.1 | - | 2 | 1870 | ||
| AK130859.1 | - | 1 | 288 | |||
| AK310465.1 | - | 2 | 154 | |||
| AK311985.1 | - | 1 | 255 | |||
| BC021180.2 | - | 1 | 239 | 5266418 | ||
| BC070148.1 | - | 2 | 154 | 6617009 | ||
| BI827031.1 | - | 1 | 228 | |||
| DQ207367.1 | - | 1 | 239 | |||

HMGB4
Genomic Map