siRNA Details
| Gene Symbol: GPR107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136557.1 | NP_001130029.1 | 7 | 811 | ||
| NM_001136558.1 | NP_001130030.1 | 7 | 811 | |||
| NM_001287346.1 | NP_001274275.1 | 7 | 807 | |||
| NM_020960.4 | NP_066011.2 | 7 | 811 | |||
| XM_017014953.1 | XP_016870442.1 | 7 | 772 | |||
| XM_017014954.1 | XP_016870443.1 | 7 | 772 | |||
| GenBank mRNA | AB046844.1 | - | 7 | 582 | ||
| AF376725.1 | - | 7 | 592 | |||
| AK026244.1 | - | 7 | 596 | |||
| AK225456.1 | - | 7 | 596 | |||
| AK293092.1 | - | 7 | 592 | |||
| AL834359.1 | - | 6 | 455 | |||
| BC110518.2 | - | 7 | 592 | 40033360 | ||
| BC143655.1 | - | 7 | 607 | |||
More Information
Set Membership: |
|

GPR107
Genomic Map