siRNA Details
| Gene Symbol: ZNRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032268.4 | NP_115644.1 | 2 | 1131 | ||
| XM_017023793.1 | XP_016879282.1 | 2 | 1105 | |||
| GenBank mRNA | AF378524.1 | - | 2 | 803 | ||
| AK294434.1 | - | 2 | 803 | |||
| AL136903.1 | - | 2 | 796 | |||
| AL834440.1 | - | 2 | 455 | |||
| BC007235.1 | - | 2 | 803 | 2823677 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001168621, NM_001168623, NM_133206, AF378525, AK079338, and others.

ZNRF1
Genomic Map