siRNA Details
| Gene Symbol: GAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002048.2 | NP_002039.2 | 1 | 2555 | ||
| GenBank mRNA | L13698.1 | - | 1 | 2555 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAS1| Gene Symbol: GAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002048.2 | NP_002039.2 | 1 | 2555 | ||
| GenBank mRNA | L13698.1 | - | 1 | 2555 | ||