siRNA Details
| Gene Symbol: PELI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020651.3 | NP_065702.2 | 3, 4 | 669 | ||
| XM_011532994.2 | XP_011531296.1 | 3, 4 | 1328 | |||
| XM_011532996.2 | XP_011531298.1 | 3, 4 | 2087 | |||
| XM_017004520.1 | XP_016860009.1 | 4, 5 | 803 | |||
| GenBank mRNA | AF300987.1 | - | 2, 3 | 224 | ||
| AF302505.1 | - | 2, 3 | 4247 | |||
| AJ278859.1 | - | 2, 3 | 209 | |||
| AK312441.1 | - | 3, 4 | 618 | |||
| AY364257.1 | - | 2, 3 | 209 | |||
| BC050019.1 | - | 3, 4 | 623 | 5296990 | ||
| BC050533.1 | - | 3, 4 | 605 | |||
| BC063611.1 | - | 3, 4 | 597 | 4992160 | ||

PELI1
Genomic Map