siRNA Details
| Gene Symbol: HOXB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002145.3 | NP_002136.1 | 2 | 1437 | ||
| XM_005257275.4 | XP_005257332.1 | 2 | 1006 | |||
| GenBank mRNA | X16665.1 | - | 2 | 1395 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXB2| Gene Symbol: HOXB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002145.3 | NP_002136.1 | 2 | 1437 | ||
| XM_005257275.4 | XP_005257332.1 | 2 | 1006 | |||
| GenBank mRNA | X16665.1 | - | 2 | 1395 | ||