siRNA Details
| Gene Symbol: FCHSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033449.2 | NP_258260.1 | 2, 3 | 170 | ||
| XM_005268524.4 | XP_005268581.1 | 2, 3 | 203 | |||
| XM_006714803.3 | XP_006714866.1 | 2, 3 | 203 | |||
| XM_011537698.2 | XP_011536000.1 | 2, 3 | 203 | |||
| XM_011537700.2 | XP_011536002.1 | 2, 3 | 204 | |||
| XM_011537701.2 | XP_011536003.1 | 2, 3 | 204 | |||
| XM_017010013.1 | XP_016865502.1 | 2, 3 | 203 | |||
| GenBank mRNA | AK000007.1 | - | 2, 3 | 154 | ||
| AY217346.1 | - | 2, 3 | 170 | |||
| BC047016.1 | - | 1, 2 | 103 | 5215830 | ||

FCHSD1
Genomic Map