siRNA Details
| Gene Symbol: FAM153B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001265615.1 | NP_001252544.1 | 6, 7 | 319 | ||
| GenBank mRNA | BC028606.1 | - | 7, 8 | 560 | 5266126 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: FAM153B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001265615.1 | NP_001252544.1 | 6, 7 | 319 | ||
| GenBank mRNA | BC028606.1 | - | 7, 8 | 560 | 5266126 | |