siRNA Details
Gene Symbol: ZNF775 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC015152.2 | - | 2 | 188 | 3546283 | |
DB472309.1 | - | 3 | 252 |
Gene Symbol: REPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL046625.1 | - | 3 | 326 |
