siRNA Details
| Gene Symbol: ZCCHC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001312891.1 | NP_001299820.1 | 3 | 993 | ||
| NM_173798.3 | NP_776159.1 | 4 | 1045 | |||
| GenBank mRNA | AK122676.1 | - | 4 | 1005 | ||
| BC031241.1 | - | 4 | 964 | 5263970 | ||
| BC036572.2 | - | 4 | 998 | 5274563 | ||
| BC057841.1 | - | 4 | 964 | 5264353 | ||

ZCCHC12
Genomic Map