siRNA Details
| Gene Symbol: PDZD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173806.3 | NP_776167.2 | 2 | 272 | ||
| NR_033694.1 | 4 | 523 | ||||
| XM_006721029.3 | XP_006721092.1 | 3 | 476 | |||
| XM_011545785.2 | XP_011544087.1 | 3 | 479 | |||
| XM_017023109.1 | XP_016878598.1 | 2 | 298 | |||
| XM_017023110.1 | XP_016878599.1 | 4 | 537 | |||
| GenBank mRNA | BC021724.1 | - | 4 | 528 | 4829370 | |
| BC028605.1 | - | 4 | 528 | 5265642 | ||
| BC039562.1 | - | 4 | 520 | 5169242 | ||
| DB450393.1 | - | 2 | 259 | |||

PDZD9
Genomic Map