siRNA Details
Gene Symbol: RNF151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174903.4 | NP_777563.2 | 3 | 732 | ||
XM_005255129.4 | XP_005255186.1 | 4 | 994 | |||
GenBank mRNA | BC029501.2 | - | 3 | 721 | ||
BC113014.2 | - | 3 | 732 | 40029536 |
