siRNA Details
| Gene Symbol: SLC26A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193476.1 | NP_001180405.1 | 2 | 218 | ||
| NM_052961.3 | NP_443193.1 | 2 | 242 | |||
| NM_138718.2 | NP_619732.2 | 2 | 280 | |||
| XM_011514294.2 | XP_011512596.1 | 2 | 461 | |||
| XM_017010235.1 | XP_016865724.1 | 2 | 268 | |||
| GenBank mRNA | AF314959.1 | - | 1 | 112 | ||
| AF331522.1 | - | 2 | 186 | |||
| AK122928.1 | - | 2 | 218 | |||
| AK125189.1 | - | 2 | 462 | |||
| BC025408.2 | - | 2 | 273 | 4823098 | ||
| BG702985.1 | - | 2 | 209 | |||

SLC26A8
Genomic Map