siRNA Details
| Gene Symbol: IL4I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258017.1 | NP_001244946.1 | 8 | 1216 | ||
| NM_001258018.1 | NP_001244947.1 | 8 | 1298 | |||
| NM_152899.1 | NP_690863.1 | 6 | 689 | |||
| NM_172374.2 | NP_758962.1 | 8 | 1240 | |||
| NR_047577.1 | 9 | 1275 | ||||
| GenBank mRNA | AF293462.1 | - | 6 | 689 | ||
| AJ880386.1 | - | 8 | 1115 | |||
| AJ880387.1 | - | 8 | 1092 | |||
| AJ880388.1 | - | 8 | 1154 | |||
| AJ880389.1 | - | 9 | 1152 | |||
| AY358933.1 | - | 6 | 689 | |||
| BC026103.1 | - | 8 | 1144 | |||
| BC064378.1 | - | 6 | 675 | 5725335 | ||
| BC090852.1 | - | 6 | 676 | 6191892 | ||
| BC131625.1 | - | 8 | 1129 | 40117891 | ||
| DQ079587.1 | - | 8 | 1240 | |||
| DQ079588.1 | - | 8 | 1298 | |||
| DQ079589.1 | - | 8 | 1216 | |||

IL4I1
Genomic Map