siRNA Details
| Gene Symbol: MORC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320445.1 | NP_001307374.1 | 14 | 2425 | ||
| NM_001320446.1 | NP_001307375.1 | 16 | 2584 | |||
| NM_015358.2 | NP_056173.1 | 15 | 2498 | |||
| XM_011529518.2 | XP_011527820.1 | 15 | 2718 | |||
| XM_017028311.1 | XP_016883800.1 | 15 | 2520 | |||
| XM_017028312.1 | XP_016883801.1 | 14 | 2343 | |||
| GenBank mRNA | AK025327.1 | - | 1 | 365 | ||
| AK292957.1 | - | 15 | 2449 | |||
| AK295133.1 | - | 14 | 2417 | |||
| BC042609.1 | - | 1 | 357 | |||
| BC094779.1 | - | 15 | 2455 | 30527767 | ||
| BC132731.1 | - | 15 | 2449 | 40146753 | ||
| BC144116.1 | - | 16 | 2538 | |||
| D50926.2 | - | 15 | 2457 | |||

MORC3
Genomic Map