siRNA Details
| Gene Symbol: ATCAY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033064.4 | NP_149053.1 | 4 | 639 | ||
| GenBank mRNA | AB058775.1 | - | 4 | 585 | ||
| AK027889.1 | - | 4 | 574 | |||
| AK092309.1 | - | 4 | 438 | |||
| AK125457.1 | - | 4 | 282 | |||
| AK299522.1 | - | 5 | 645 | |||
| AY220297.1 | - | 3 | 209 | |||
| BC026217.1 | - | 4 | 581 | 4153341 | ||
| KU178814.1 | - | 3 | 209 | |||
| KU178815.1 | - | 3 | 209 | |||

ATCAY
Genomic Map