siRNA Details
| Gene Symbol: SLC26A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193476.1 | NP_001180405.1 | 10 | 1327 | ||
| NM_052961.3 | NP_443193.1 | 10 | 1351 | |||
| NM_138718.2 | NP_619732.2 | 8 | 1074 | |||
| XM_011514294.2 | XP_011512596.1 | 10 | 1570 | |||
| XM_017010235.1 | XP_016865724.1 | 10 | 1377 | |||
| GenBank mRNA | AF314959.1 | - | 9 | 1221 | ||
| AF331522.1 | - | 10 | 1295 | |||
| AK097410.1 | - | 4 | 413 | |||
| AK122928.1 | - | 10 | 1327 | |||
| AK125189.1 | - | 10 | 1571 | |||
| BC025408.2 | - | 8 | 1067 | 4823098 | ||

SLC26A8
Genomic Map