siRNA Details
| Gene Symbol: CCDC116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152612.2 | NP_689825.2 | 4 | 923 | ||
| XM_006724159.2 | XP_006724222.1 | 4 | 1142 | |||
| XM_011529984.2 | XP_011528286.1 | 3 | 1465 | |||
| GenBank mRNA | AK093365.1 | - | 4 | 991 | ||
| AK302030.1 | - | 4 | 1004 | |||
| BC033499.1 | - | 4 | 923 | 5172053 | ||

CCDC116
Genomic Map