siRNA Details
| Gene Symbol: ZNF792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175872.4 | NP_787068.3 | 4 | 2044 | ||
| GenBank mRNA | AK095770.1 | - | 1 | 436 | ||
| AK130901.1 | - | 1 | 753 | |||
| AK304550.1 | - | 4 | 1959 | |||
| BC101118.1 | - | 2 | 1440 | 40027074 | ||
| BC101119.2 | - | 2 | 1440 | 40027075 | ||
| BC101120.1 | - | 2 | 1440 | 40027081 | ||

ZNF792
Genomic Map