siRNA Details
| Gene Symbol: C12orf60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175874.3 | NP_787070.2 | 2 | 416 | ||
| XM_005253322.4 | XP_005253379.1 | 2 | 688 | |||
| XM_011520568.2 | XP_011518870.1 | 2 | 1084 | |||
| XM_011520569.2 | XP_011518871.1 | 2 | 260 | |||
| XM_017018871.1 | XP_016874360.1 | 7 | 1127 | |||
| XM_017018872.1 | XP_016874361.1 | 6 | 554 | |||
| XM_017018873.1 | XP_016874362.1 | 2 | 419 | |||
| XM_017018874.1 | XP_016874363.1 | 2 | 259 | |||
| GenBank mRNA | AK289942.1 | - | 2 | 313 | ||
| BC015546.1 | - | 2 | 288 | 3916539 | ||
| BC029443.1 | - | 2 | 273 | 4670900 | ||
| BC038836.1 | - | 2 | 416 | 5168757 | ||

C12orf60
Genomic Map