siRNA Details
| Gene Symbol: ZADH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001306093.1 | NP_001293022.1 | 2 | 637 | ||
| NM_175907.5 | NP_787103.1 | 2 | 815 | |||
| GenBank mRNA | AK292880.1 | - | 2 | 570 | ||
| AK302801.1 | - | 2 | 367 | |||
| BC018081.1 | - | 2 | 793 | |||
| BC033780.1 | - | 2 | 701 | 4510603 | ||
| BC078661.1 | - | 2 | 628 | 5575156 | ||
| BX361234.2 | - | 2 | 817 | |||

ZADH2
Genomic Map