siRNA Details
| Gene Symbol: C12orf60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175874.3 | NP_787070.2 | 2 | 788 | ||
| XM_005253322.4 | XP_005253379.1 | 2 | 1060 | |||
| XM_011520568.2 | XP_011518870.1 | 2 | 1456 | |||
| XM_011520569.2 | XP_011518871.1 | 2 | 632 | |||
| XM_017018871.1 | XP_016874360.1 | 7 | 1499 | |||
| XM_017018872.1 | XP_016874361.1 | 6 | 926 | |||
| XM_017018873.1 | XP_016874362.1 | 2 | 791 | |||
| XM_017018874.1 | XP_016874363.1 | 2 | 631 | |||
| GenBank mRNA | AK289942.1 | - | 2 | 685 | ||
| BC015546.1 | - | 2 | 660 | 3916539 | ||
| BC029443.1 | - | 2 | 645 | 4670900 | ||
| BC038836.1 | - | 2 | 788 | 5168757 | ||

C12orf60
Genomic Map