siRNA Details
| Gene Symbol: ZADH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001306093.1 | NP_001293022.1 | 2 | 705 | ||
| NM_175907.5 | NP_787103.1 | 2 | 883 | |||
| GenBank mRNA | AK292880.1 | - | 2 | 638 | ||
| AK302801.1 | - | 2 | 435 | |||
| BC018081.1 | - | 2 | 861 | |||
| BC033780.1 | - | 2 | 769 | 4510603 | ||
| BC078661.1 | - | 2 | 696 | 5575156 | ||
| BX647730.1 | - | 2 | 707 | |||

ZADH2
Genomic Map