siRNA Details
| Gene Symbol: RAB7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164522.2 | NP_001157994.1 | 5, 6 | 759 | ||
| NM_177403.5 | NP_796377.3 | 4, 5 | 833 | |||
| GenBank mRNA | AK127140.1 | - | 4, 5 | 817 | ||
| AY094596.1 | - | 5, 6 | 706 | |||
| BC017092.1 | - | 5, 6 | 701 | 3851998 | ||

RAB7B
Genomic Map