siRNA Details
| Gene Symbol: TRAPPC6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079537.1 | NP_001073005.1 | 2, 3 | 492 | ||
| NM_177452.3 | NP_803235.1 | 2, 3 | 492 | |||
| XM_011536409.2 | XP_011534711.1 | 3, 4 | 512 | |||
| XM_017020964.1 | XP_016876453.1 | 2, 3 | 465 | |||
| GenBank mRNA | AK056690.1 | - | 2, 3 | 462 | ||
| AL833179.1 | - | 2, 3 | 376 | |||
| BC047328.1 | - | 2, 3 | 376 | 5534151 | ||
| BX161510.1 | - | 2, 3 | 384 | |||

TRAPPC6B
Genomic Map