siRNA Details
| Gene Symbol: C8orf37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177965.3 | NP_808880.1 | 5, 6 | 523 | ||
| GenBank mRNA | AK055162.1 | - | 5, 6 | 498 | ||
| BC036855.2 | - | 5, 6 | 488 | |||
| DB205618.1 | - | 5, 6 | 488 | |||
| GQ240139.1 | - | 5, 6 | 498 | |||

C8orf37
Genomic Map