siRNA Details
| Gene Symbol: PBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204961.1 | NP_001191890.1 | 6 | 1371 | ||
| NM_001204963.1 | NP_001191892.1 | 6 | 1371 | |||
| NM_002585.3 | NP_002576.1 | 6 | 1371 | |||
| XM_005245228.3 | XP_005245285.1 | 7 | 1539 | |||
| XM_005245229.3 | XP_005245286.1 | 7 | 1132 | |||
| XM_011509590.1 | XP_011507892.1 | 7 | 1539 | |||
| XM_011509591.2 | XP_011507893.1 | 7 | 1539 | |||
| XM_011509592.1 | XP_011507894.1 | 7 | 1539 | |||
| XM_017001395.1 | XP_016856884.1 | 6 | 1371 | |||
| XM_017001396.1 | XP_016856885.1 | 6 | 1371 | |||
| GenBank mRNA | AK291415.1 | - | 6 | 1053 | ||
| AK299673.1 | - | 6 | 987 | |||
| AK301540.1 | - | 5 | 716 | |||
| BC101578.1 | - | 6 | 960 | 8069084 | ||
| BC143435.1 | - | 5 | 886 | |||
| BC143451.1 | - | 6 | 960 | |||
| BT006705.1 | - | 6 | 908 | |||
| M31170.1 | - | 4 | 644 | |||
| M86546.1 | - | 6 | 1031 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_183355, NM_001291508, NM_008783, BC058390, AB221563, and others.

PBX1
Genomic Map