siRNA Details
| Gene Symbol: LRRC20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278211.1 | NP_001265140.1 | 5 | 671 | ||
| NM_001278212.1 | NP_001265141.1 | 5 | 616 | |||
| NM_001278213.1 | NP_001265142.1 | 4 | 415 | |||
| NM_001278214.1 | NP_001265143.1 | 4 | 471 | |||
| NM_018205.3 | NP_060675.1 | 4 | 448 | |||
| NM_018239.3 | NP_060709.2 | 4 | 466 | |||
| NM_207119.2 | NP_997002.1 | 5 | 929 | |||
| NR_103467.1 | 3 | 321 | ||||
| NR_103468.1 | 3 | 303 | ||||
| XM_017016374.1 | XP_016871863.1 | 5 | 611 | |||
| XM_017016375.1 | XP_016871864.1 | 5 | 606 | |||
| GenBank mRNA | AK001613.1 | - | 4 | 424 | ||
| AK001706.1 | - | 4 | 442 | |||
| AK094734.1 | - | 5 | 612 | |||
| AL122117.1 | - | 2 | 153 | |||
| AY358318.1 | - | 5 | 673 | |||
| BC024001.2 | - | 5 | 607 | 4822655 | ||
| BM788766.1 | - | 5 | 590 | |||
| CF552977.1 | - | 5 | 585 | |||
| DA032832.1 | - | 3 | 295 | |||
| DA160782.1 | - | 4 | 447 | |||
| DA386385.1 | - | 3 | 277 | |||
| DA704994.1 | - | 4 | 415 | |||

LRRC20
Genomic Map