siRNA Details
| Gene Symbol: PDGFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025208.4 | NP_079484.1 | 2 | 551 | ||
| NM_033135.3 | NP_149126.1 | 2 | 533 | |||
| GenBank mRNA | AB033832.1 | - | 2 | 335 | ||
| AF113216.1 | - | 2 | 285 | |||
| AF335584.1 | - | 2 | 360 | |||
| AF336376.1 | - | 2 | 354 | |||
| AK292801.1 | - | 2 | 359 | |||
| AY027517.1 | - | 2 | 271 | |||
| AY027518.1 | - | 2 | 253 | |||
| AY359116.1 | - | 2 | 192 | |||
| BC030645.2 | - | 2 | 613 | 4824526 | ||

PDGFD
Genomic Map