siRNA Details
Gene Symbol: MAGED4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242362.1 | NP_001229291.1 | 2 | 284 | ||
NM_030801.3 | NP_110428.2 | 2 | 263 | |||
NM_177535.2 | NP_803879.1 | 2 | 233 | |||
NM_177537.2 | NP_803881.1 | 2 | 233 | |||
XM_006724607.3 | XP_006724670.1 | 2 | 249 | |||
XM_006724608.3 | XP_006724671.1 | 2 | 275 | |||
XM_011530822.1 | XP_011529124.1 | 2 | 190 | |||
XM_011530823.2 | XP_011529125.1 | 2 | 190 | |||
XM_017029876.1 | XP_016885365.1 | 2 | 249 | |||
GenBank mRNA | AB040527.1 | - | 2 | 144 | ||
AB040528.1 | - | 2 | 144 | |||
AB040529.1 | - | 2 | 234 | |||
AB058762.2 | - | 2 | 239 | |||
AK098830.1 | - | 2 | 219 | |||
AK289458.1 | - | 2 | 198 | |||
AK308365.1 | - | 2 | 198 | |||
AL136582.1 | - | 2 | 182 | |||
BC001207.1 | - | 2 | 231 | |||
BC063838.1 | - | 2 | 145 | 6185437 | ||
BC081566.1 | - | 2 | 157 | 6292139 |
Gene Symbol: MAGED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098800.2 | NP_001092270.1 | 2 | 233 | ||
NM_001272061.1 | NP_001258990.1 | 2 | 284 | |||
NM_001272062.1 | NP_001258991.1 | 2 | 233 | |||
NM_001272063.1 | NP_001258992.1 | 2 | 263 |
