siRNA Details
| Gene Symbol: TBRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261834.1 | NP_001248763.1 | 3 | 653 | ||
| NM_004749.3 | NP_004740.2 | 3 | 697 | |||
| NM_030900.3 | NP_112162.1 | 3 | 697 | |||
| NM_199122.2 | NP_954573.1 | 3 | 676 | |||
| GenBank mRNA | AB023165.1 | - | 3 | 668 | ||
| AF011792.1 | - | 1 | 36 | |||
| AK001281.1 | - | 3 | 645 | |||
| AK022575.1 | - | 3 | 633 | |||
| AK092132.1 | - | 1 | 1831 | |||
| AK222943.1 | - | 3 | 633 | |||
| AK298155.1 | - | 1 | 46 | |||
| AK300489.1 | - | 3 | 612 | |||
| AL833840.1 | - | 3 | 666 | |||
| BC002732.2 | - | 1 | 136 | |||
| BC014918.1 | - | 3 | 627 | 3906670 | ||
| BC017235.2 | - | 3 | 624 | 3538504 | ||
| BG703429.1 | - | 3 | 662 | |||

TBRG4
Genomic Map