siRNA Details
| Gene Symbol: RRAGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006064.4 | NP_006055.3 | 3 | 805 | ||
| NM_016656.3 | NP_057740.2 | 3 | 805 | |||
| XM_005261973.4 | XP_005262030.1 | 3 | 142 | |||
| XM_017029217.1 | XP_016884706.1 | 3 | 135 | |||
| XM_017029218.1 | XP_016884707.1 | 3 | 761 | |||
| GenBank mRNA | AK289407.1 | - | 3 | 738 | ||
| AL831926.1 | - | 3 | 744 | |||
| BC034726.1 | - | 3 | 736 | 3919252 | ||
| X90530.1 | - | 3 | 596 | |||

RRAGB
Genomic Map