siRNA Details
| Gene Symbol: ATG16L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190266.1 | NP_001177195.1 | 11 | 1365 | ||
| NM_001190267.1 | NP_001177196.1 | 11 | 1363 | |||
| NM_017974.3 | NP_060444.3 | 10 | 1310 | |||
| NM_030803.6 | NP_110430.5 | 11 | 1367 | |||
| NM_198890.2 | NP_942593.2 | 7 | 878 | |||
| XM_005246082.2 | XP_005246139.1 | 12 | 1355 | |||
| XM_005246084.2 | XP_005246141.1 | 9 | 894 | |||
| XM_005246086.2 | XP_005246143.1 | 8 | 843 | |||
| XM_006712608.2 | XP_006712671.1 | 11 | 1166 | |||
| GenBank mRNA | AK000897.1 | - | 10 | 978 | ||
| AK001690.1 | - | 10 | 1155 | |||
| AK024453.1 | - | 10 | 1905 | |||
| AK027854.1 | - | 9 | 892 | |||
| AK074434.1 | - | 5 | 337 | |||
| AK123876.1 | - | 7, 8 | 729 | |||
| AK308400.1 | - | 10 | 1208 | |||
| AY358182.1 | - | 11 | 1256 | |||
| AY398617.1 | - | 11 | 1256 | |||
| BC071846.1 | - | 7 | 716 | |||
| BC117337.1 | - | 11 | 1029 | |||
| BC143900.1 | - | 10 | 972 | |||
| EF079889.1 | - | 10 | 1053 | |||
| EF079890.1 | - | 7 | 621 | |||
| EF560734.1 | - | 10 | 1180 | |||
| JQ924062.1 | - | 10 | 1053 | |||

ATG16L1
Genomic Map