siRNA Details
| Gene Symbol: PDGFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025208.4 | NP_079484.1 | 2 | 686 | ||
| NM_033135.3 | NP_149126.1 | 2 | 668 | |||
| GenBank mRNA | AB033832.1 | - | 2 | 470 | ||
| AF113216.1 | - | 2 | 420 | |||
| AF335584.1 | - | 2 | 495 | |||
| AF336376.1 | - | 2 | 489 | |||
| AK292801.1 | - | 2 | 494 | |||
| AY027517.1 | - | 2 | 406 | |||
| AY027518.1 | - | 2 | 388 | |||
| AY359116.1 | - | 2 | 327 | |||
| BC030645.2 | - | 2 | 748 | 4824526 | ||
| BG722948.1 | - | 2 | 558 | |||

PDGFD
Genomic Map