siRNA Details
| Gene Symbol: NDUFAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144736.4 | NP_653337.1 | 6 | 712 | ||
| XM_005264410.1 | XP_005264467.1 | 6 | 692 | |||
| XM_011532956.1 | XP_011531258.1 | 6 | 648 | |||
| GenBank mRNA | AK302150.1 | - | 5 | 582 | ||
| BC004548.2 | - | 6 | 677 | 3959139 | ||
| BC012374.1 | - | 1 | 26 | |||
| BI334629.1 | - | 6 | 509 | |||

NDUFAF7
Genomic Map