siRNA Details
| Gene Symbol: CSAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102576.2 | NP_001096046.2 | 3 | 195 | ||
| NM_153478.2 | NP_705611.2 | 4 | 411 | |||
| GenBank mRNA | AF268418.1 | - | 3 | 165 | ||
| AF268419.1 | - | 4 | 362 | |||
| BC059947.1 | - | 3 | 165 | 5397842 | ||
| BF570999.1 | - | 3 | 194 | |||
| BM810082.1 | - | 3 | 177 | |||

CSAG1
Genomic Map