siRNA Details
| Gene Symbol: SIX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175875.4 | NP_787071.2 | 3 | 2561 | ||
| GenBank mRNA | BC033204.1 | - | 2 | 632 | ||
| BC084563.1 | - | 1 | 262 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SIX5| Gene Symbol: SIX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175875.4 | NP_787071.2 | 3 | 2561 | ||
| GenBank mRNA | BC033204.1 | - | 2 | 632 | ||
| BC084563.1 | - | 1 | 262 | |||