siRNA Details
| Gene Symbol: SYVN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032431.2 | NP_115807.1 | 3 | 279 | ||
| NM_172230.2 | NP_757385.1 | 3 | 279 | |||
| XM_011545303.2 | XP_011543605.1 | 3 | 1127 | |||
| GenBank mRNA | AB058713.1 | - | 3 | 227 | ||
| AB085847.1 | - | 2 | 185 | |||
| AF317634.1 | - | 2 | 185 | |||
| AK074148.1 | - | 3 | 234 | |||
| AK126912.1 | - | 3 | 280 | |||
| AL834262.1 | - | 3 | 279 | |||
| BC030530.1 | - | 3 | 220 | 5211742 | ||

SYVN1
Genomic Map