siRNA Details
| Gene Symbol: ZNF493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076678.2 | NP_001070146.1 | 4 | 2372 | ||
| NM_175910.6 | NP_787106.4 | 2 | 2149 | |||
| GenBank mRNA | AB209086.1 | - | 2 | 2117 | ||
| AK093823.1 | - | 1 | 742 | |||
| AK095616.1 | - | 1 | 901 | |||
| AK127476.1 | - | 1 | 1757 | |||
| AK128494.1 | - | 2 | 2135 | |||
| BC112346.1 | - | 1 | 1968 | |||
| BX648842.1 | - | 4 | 2362 | |||

ZNF493
Genomic Map