siRNA Details
| Gene Symbol: PPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006903.4 | NP_008834.3 | 8 | 770 | ||
| NM_176866.2 | NP_789842.2 | 5 | 551 | |||
| NM_176867.3 | NP_789843.2 | 3 | 359 | |||
| NM_176869.2 | NP_789845.1 | 9 | 857 | |||
| GenBank mRNA | AF217187.1 | - | 9 | 851 | ||
| AK289607.1 | - | 8 | 748 | |||
| BC022803.1 | - | 3 | 335 | |||
| BC035569.1 | - | 9 | 829 | |||
| BC039462.1 | - | 5 | 498 | |||
| BC057219.1 | - | 8 | 763 | 4289401 | ||

PPA2
Genomic Map