siRNA Details
| Gene Symbol: MIDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177401.4 | NP_796375.3 | 8 | 2285 | ||
| XM_005259671.3 | XP_005259728.1 | 9 | 2414 | |||
| XM_005259672.3 | XP_005259729.1 | 9 | 2412 | |||
| GenBank mRNA | AK075506.1 | - | 1 | 390 | ||
| BC015089.2 | - | 2 | 895 | |||
| BC094778.1 | - | 7 | 2182 | 30342357 | ||

MIDN
Genomic Map