siRNA Details
| Gene Symbol: SULT1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001054.3 | NP_001045.1 | 8 | 1027 | ||
| NM_177528.2 | NP_803564.1 | 8 | 1300 | |||
| XM_006721077.3 | XP_006721140.1 | 4 | 1564 | |||
| XM_017023600.1 | XP_016879089.1 | 8 | 1054 | |||
| XM_017023601.1 | XP_016879090.1 | 8 | 1482 | |||
| XM_017023602.1 | XP_016879091.1 | 9 | 1184 | |||
| XM_017023603.1 | XP_016879092.1 | 6 | 810 | |||
| GenBank mRNA | U28169.1 | - | 8 | 1022 | ||
| U28170.1 | - | 8 | 1010 | |||
| X78282.1 | - | 7 | 949 | |||

SULT1A2
Genomic Map