siRNA Details
| Gene Symbol: TMEM158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015444.2 | NP_056259.2 | 1 | 1356 | ||
| GenBank mRNA | AF438313.1 | - | 1 | 1296 | ||
| AL080235.1 | - | 1 | 611 | |||
| BC057390.1 | - | 2 | 1270 | 6175193 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM158| Gene Symbol: TMEM158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015444.2 | NP_056259.2 | 1 | 1356 | ||
| GenBank mRNA | AF438313.1 | - | 1 | 1296 | ||
| AL080235.1 | - | 1 | 611 | |||
| BC057390.1 | - | 2 | 1270 | 6175193 | ||