siRNA Details
| Gene Symbol: HS1BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022460.3 | NP_071905.3 | 7 | 1365 | ||
| XM_017004700.1 | XP_016860189.1 | 7 | 1464 | |||
| GenBank mRNA | AK024311.1 | - | 6 | 777 | ||
| BC050636.1 | - | 7 | 1345 | 5193161 | ||
| BG753647.1 | - | 4 | 758 | |||

HS1BP3
Genomic Map