siRNA Details
| Gene Symbol: GPR62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080865.3 | NP_543141.3 | 1 | 1439 | ||
| GenBank mRNA | AB032600.1 | - | 1 | 1405 | ||
| BC026357.1 | - | 1 | 1372 | 4812283 | ||
More Information
Set Membership: |
|

GPR62
Genomic Map