siRNA Details
| Gene Symbol: MCRIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138418.2 | NP_612427.2 | 5 | 615 | ||
| GenBank mRNA | AK054811.1 | - | 3 | 2011 | ||
| BC001912.1 | - | 5 | 615 | 3546959 | ||
| BC007346.2 | - | 4 | 407 | |||
| BC111391.1 | - | 5 | 517 | 6457301 | ||

MCRIP2
Genomic Map